Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009752.2 | NP_033882.1 | 7 | 886 | ||
GenBank mRNA | AK034049.1 | - | 7 | 865 | ||
AK034966.1 | - | 687 | ||||
AK042069.1 | - | 7 | 856 | |||
AK048460.1 | - | 7 | 817 | |||
AK076517.1 | - | 7 | 857 | |||
AK153218.1 | - | 7 | 868 | |||
AK167271.1 | - | 5 | 496 | |||
AK171593.1 | - | 7 | 889 | |||
AK171760.1 | - | 6 | 687 | |||
BC028875.1 | - | 7 | 806 | 4240285 | ||
M57734.1 | - | 7 | 845 |