Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110504.1 | NP_001103974.1 | 14, 15 | 1694 | ||
NM_007600.3 | NP_031626.1 | 14, 15 | 1746 | |||
GenBank mRNA | AF021847.1 | - | 14, 15 | 1625 | ||
AF084459.1 | - | 13, 14 | 1612 | |||
AK088547.1 | - | 14, 15 | 1666 | |||
AK149090.1 | - | 10, 11 | 1094 | |||
AK149706.1 | - | 14, 15 | 1687 | |||
AK150084.1 | - | 14, 15 | 1685 | |||
AK160940.1 | - | 14, 15 | 1686 | |||
AK165258.1 | - | 14, 15 | 1706 | |||
AK168063.1 | - | 14, 15 | 1685 | |||
AK171405.1 | - | 14, 15 | 1649 | |||
BC026138.1 | - | 14, 15 | 1661 | 4218148 | ||
BC050276.1 | - | 14, 15 | 1662 | 4923347 |