Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037761.2 | NP_001032850.1 | 4 | 361 | ||
NM_001271405.1 | NP_001258334.1 | 4 | 340 | |||
NM_001271406.1 | NP_001258335.1 | 4 | 326 | |||
NM_009798.4 | NP_033928.1 | 4 | 361 | |||
GenBank mRNA | AK153367.1 | - | 4 | 292 | ||
AK153394.1 | - | 4 | 292 | |||
AK156778.1 | - | 4 | 330 | |||
AK159390.1 | - | 4 | 335 | |||
AK160082.1 | - | 4 | 336 | |||
AK162757.1 | - | 4 | 328 | |||
AK168738.1 | - | 4 | 289 | |||
BC002053.1 | - | 4 | 303 | 3590980 | ||
BY033332.1 | - | 4 | 360 | |||
FJ692320.1 | - | 4 | 326 | |||
U10406.1 | - | 4 | 281 | |||
U10407.1 | - | 4 | 252 |