Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009931.2 | NP_034061.2 | 24, 25 | 1758 | ||
GenBank mRNA | AK146487.1 | - | 24, 25 | 1707 | ||
AK147284.1 | - | 24, 25 | 1680 | |||
AK147355.1 | - | 24, 25 | 1683 | |||
AK147661.1 | - | 24, 25 | 1724 | |||
AK159130.1 | - | 24, 25 | 1710 | |||
BC072650.1 | - | 24, 25 | 1621 | 6852661 | ||
J04694.1 | - | 24, 25 | 1662 | |||
X06777.1 | - | 24, 25 | 1681 |