Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 37 | 2424 | ||
GenBank mRNA | AK075707.1 | - | 15 | 995 | ||
AK132496.1 | - | 37 | 2420 | |||
AK159212.1 | - | 37 | 2425 | |||
AK159408.1 | - | 37 | 2421 | |||
AK159578.1 | - | 37 | 2422 | |||
AK159958.1 | - | 16 | 1043 | |||
AK160098.1 | - | 37 | 2424 | |||
AK160787.1 | - | 37 | 2420 | |||
AK160788.1 | - | 37 | 2419 | |||
AK160790.1 | - | 37 | 2420 | |||
AK160814.1 | - | 29 | 1889 | |||
AK160946.1 | - | 37 | 2423 | |||
AK161017.1 | - | 37 | 2425 | |||
AK161605.1 | - | 37 | 2420 | |||
AK164628.1 | - | 16 | 1043 | |||
AK164808.1 | - | 37 | 2420 | |||
BC007158.1 | - | 37 | 2404 | 3487477 | ||
BC042503.1 | - | 37 | 2412 | 3586272 | ||
X58251.1 | - | 37 | 2360 |