Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007747.2 | NP_031773.2 | 3 | 375 | ||
GenBank mRNA | AK018723.1 | - | 3 | 310 | ||
BC003831.1 | - | 2 | 1501 | |||
BC034302.1 | - | 3 | 328 | 5372849 | ||
BY259203.1 | - | 3 | 377 | |||
CO044753.1 | - | 3 | 330 | |||
X15963.1 | - | 3 | 299 |