Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021282.2 | NP_067257.1 | 6 | 897 | ||
GenBank mRNA | BC013451.1 | - | 6 | 897 | 4223797 | |
BC042693.1 | - | 6 | 876 | |||
L11650.1 | - | 6 | 990 | |||
X62595.1 | - | 6 | 882 |