Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007854.3 | NP_031880.2 | 4, 5 | 644 | ||
GenBank mRNA | AF183397.1 | - | 4, 5 | 554 | ||
AK010950.1 | - | 4, 5 | 581 | |||
AK083484.1 | - | 4, 5 | 541 | |||
AK144451.1 | - | 4, 5 | 537 | |||
AK154310.1 | - | 4, 5 | 579 | |||
BC048958.1 | - | 4, 5 | 790 | 6491511 | ||
X86682.1 | - | 2, 3 | 212 |