Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034996.1 | - | 5 | 1677 | ||
BC006797.1 | - | 5 | 1646 | 2648527 | ||
BC018572.1 | - | 5 | 1487 | |||
BC021656.1 | - | 5 | 1659 | 3602195 | ||
U12983.1 | - | 5 | 1174 | |||
Z48781.1 | - | 5 | 1637 |