Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007987.2 | NP_032013.2 | 8, 9 | 712 | ||
GenBank mRNA | AK002590.1 | - | 8, 9 | 663 | ||
BC061160.1 | - | 8, 9 | 685 | 30302649 | ||
DQ846748.1 | - | 8, 9 | 656 | |||
DQ846749.1 | - | 8, 9 | 656 | |||
M83649.1 | - | 8, 9 | 705 |