Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fhl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010214.4 | NP_034344.2 | 2 | 1079 | ||
GenBank mRNA | AF053486.2 | - | 2 | 1072 | ||
AK005848.1 | - | 2 | 1050 | |||
AK006464.1 | - | 2 | 1055 | |||
AK029585.1 | - | 1081 | ||||
AK030256.1 | - | 2 | 992 | |||
AK161287.1 | - | 2 | 1057 | |||
AK161369.1 | - | 2 | 1232 | |||
BC046583.1 | - | 1 | 810 | 6492801 | ||
BC061148.1 | - | 2 | 1026 | 6774577 |