Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aktip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146459.1 | - | 10 | 995 | ||
AK158704.1 | - | 10 | 997 | |||
BC008130.1 | - | 11 | 1000 | 3498984 | ||
X71978.1 | - | 10 | 1019 | |||
Z67963.1 | - | 10 | 1019 |