Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6pd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019468.2 | NP_062341.2 | 1 | 266 | ||
GenBank mRNA | BC120827.1 | - | 1 | 266 | 40129750 | |
BC137684.1 | - | 1 | 266 | 8860703 |