Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008093.2 | NP_032119.2 | 1, 2 | 853 | ||
GenBank mRNA | AK132020.1 | - | 1, 2 | 874 | ||
AK142213.1 | - | 1, 2 | 855 | |||
BC105653.1 | - | 2, 3 | 566 | 40046623 | ||
BC105654.1 | - | 2, 3 | 627 | 40046622 | ||
BC105655.1 | - | 2, 3 | 476 | 40046621 | ||
BC105656.1 | - | 2, 3 | 132 | |||
U84725.1 | - | 1, 2 | 847 |