Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267621.1 | NP_001254550.1 | 7 | 1548 | ||
NM_010278.2 | NP_034408.1 | 6 | 1760 | |||
GenBank mRNA | AK089065.1 | - | 7 | 1549 | ||
AK132814.1 | - | 1 | 416 | |||
BC130219.1 | - | 5 | 945 | |||
BC152328.1 | - | 5 | 836 | |||
BU698904.1 | - | 4 | 576 | |||
U58972.1 | - | 6 | 1735 | |||
U78312.1 | - | 6 | 1760 |