Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008154.3 | NP_032180.1 | 2 | 560 | ||
GenBank mRNA | AK140302.1 | - | 2 | 531 | ||
AK140464.1 | - | 2 | 514 | |||
AK158917.1 | - | 2 | 523 | |||
BC138248.1 | - | 1 | 882 | 8861267 | ||
BC138250.1 | - | 1 | 882 | 8861269 | ||
D21062.1 | - | 1 | 935 |
Set Membership: |
G-protein coupled receptor |