Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gstm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008184.3 | NP_032210.3 | 2 | 178 | ||
GenBank mRNA | AJ000413.1 | - | 2 | 62 | ||
AK015265.1 | - | 2 | 164 | |||
BC031818.1 | - | 2 | 141 | 4196506 | ||
CB573365.1 | - | 2 | 181 |