Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010662.1 | NP_034792.1 | 1 | 86 | ||
GenBank mRNA | AK004635.1 | - | 1 | 158 | ||
AK010156.1 | - | 1 | 158 | |||
AK132640.1 | - | 1 | 158 | |||
BC060286.1 | - | 1 | 101 | 30292772 |