Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162432.1 | NP_001155904.1 | 12 | 1306 | ||
NM_001162433.1 | NP_001155905.1 | 12 | 1348 | |||
NM_010693.3 | NP_034823.1 | 11 | 1403 | |||
GenBank mRNA | AK088001.1 | - | 11 | 1404 | ||
AK166263.1 | - | 6 | 793 | |||
BC011474.1 | - | 12 | 1313 | 4191610 | ||
CX217876.1 | - | 4 | 562 | |||
M12056.1 | - | 11 | 1379 | |||
X03533.1 | - | 11 | 1409 |