Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281980.1 | NP_001268909.1 | 9 | 697 | ||
NM_001281981.1 | NP_001268910.1 | 10 | 738 | |||
NM_008511.3 | NP_032537.3 | 9 | 969 | |||
GenBank mRNA | AK040180.1 | - | 9 | 970 | ||
AK050646.1 | - | 9 | 1407 | |||
AK079932.1 | - | 9 | 927 | |||
AK153823.1 | - | 9 | 967 | |||
AK153865.1 | - | 9 | 699 | |||
AK160009.1 | - | 9 | 929 | |||
AK166635.1 | - | 7 | 418 | |||
AK168775.1 | - | 7 | 395 | |||
AK169578.1 | - | 10 | 722 | |||
AY753324.1 | - | 8 | 532 | |||
BC052909.1 | - | 9 | 905 | 30076553 | ||
U10484.1 | - | 9 | 675 |