Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010745.2 | NP_034875.1 | 1 | 213 | ||
GenBank mRNA | AB007599.1 | - | 1 | 213 | ||
AK027929.1 | - | 1 | 202 | |||
BC065783.1 | - | 1 | 118 | |||
BC096414.1 | - | 1 | 181 | 849991 |