Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008536.3 | NP_032562.1 | 4, 5 | 861 | ||
GenBank mRNA | AK077326.1 | - | 2, 3 | 364 | ||
AK133556.1 | - | 362 | ||||
AK134603.1 | - | 3, 4 | 797 | |||
AK164718.1 | - | 2, 3 | 365 | |||
BC083073.1 | - | 4, 5 | 861 | 6337376 | ||
KC508089.1 | - | 2, 3 | 336 | |||
L15429.1 | - | 2, 3 | 351 | |||
L15443.1 | - | 2, 3 | 351 |