Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012335.2 | NP_001012335.1 | 4 | 545 | ||
NM_001012336.2 | NP_001012336.1 | 4 | 542 | |||
NM_001291481.1 | NP_001278410.1 | 4 | 452 | |||
NM_001291482.1 | NP_001278411.1 | 3 | 465 | |||
NM_010784.5 | NP_034914.1 | 4 | 744 | |||
GenBank mRNA | AK032742.1 | - | 4 | 448 | ||
AK084457.1 | - | 4 | 465 | |||
AY864926.1 | - | 4 | 478 | |||
BC012244.1 | - | 4 | 413 | 4167496 | ||
CF728526.1 | - | 4 | 438 | |||
CF738400.1 | - | 4 | 550 | |||
CF740555.1 | - | 2 | 262 | |||
CJ174578.1 | - | 3 | 340 | |||
DQ323886.1 | - | 4 | 520 | |||
M19662.1 | - | 3 | 623 | |||
M34327.1 | - | 4 | 418 | |||
M34328.1 | - | 4 | 729 | |||
M35833.1 | - | 4 | 418 |