Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252282.1 | NP_001239211.1 | 19 | 2566 | ||
NM_001252283.1 | NP_001239212.1 | 19 | 2554 | |||
NM_001252287.1 | NP_001239216.1 | 18 | 2597 | |||
NM_001252288.1 | NP_001239217.1 | 18 | 2569 | |||
NM_010956.4 | NP_035086.2 | 18 | 2521 | |||
GenBank mRNA | AK147289.1 | - | 18 | 2522 | ||
AK150009.1 | - | 18 | 2487 | |||
AK169286.1 | - | 18 | 2513 | |||
AK220536.1 | - | 19 | 2528 | |||
BC012042.1 | - | 2 | 156 | |||
BC013670.1 | - | 18 | 2455 | 4188988 | ||
BC025040.1 | - | 3 | 321 | |||
BC029143.1 | - | 3 | 331 | |||
BC031165.1 | - | 3 | 300 | |||
BC049104.1 | - | 18 | 2522 | 6417392 | ||
BC057354.1 | - | 19 | 2482 | 6827509 |