Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008842.3 | NP_032868.2 | 4 | 368 | ||
GenBank mRNA | AK047301.1 | - | 229 | |||
AK154669.1 | - | 3 | 207 | |||
AK158445.1 | - | 4 | 620 | |||
BC042885.1 | - | 4 | 601 | 4507947 | ||
BC053019.1 | - | 4 | 368 | 5694941 | ||
BC055316.1 | - | 4 | 368 | 5694941 |