Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011125.2 | NP_035255.1 | 11, 12 | 1232 | ||
GenBank mRNA | AK148330.1 | - | 11, 12 | 1237 | ||
AK149730.1 | - | 11, 12 | 1163 | |||
BC003782.1 | - | 11, 12 | 1166 | 3491360 | ||
CT010317.1 | - | 10, 11 | 1096 | |||
U28960.1 | - | 10, 11 | 1090 | |||
U37226.1 | - | 11, 12 | 1162 |