Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008894.2 | NP_032920.2 | 5, 6 | 706 | ||
GenBank mRNA | AK013901.1 | - | 5, 6 | 673 | ||
AK181645.1 | - | 260 | ||||
BC003342.1 | - | 5, 6 | 672 | 3257610 | ||
CB205181.1 | - | 2, 3 | 242 | |||
Z72486.1 | - | 5, 6 | 667 |