Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004025.4 | NP_001004025.1 | 1 | 323 | ||
GenBank mRNA | AK029998.1 | - | 1 | 325 | ||
AK132760.1 | - | 1 | 348 | |||
AK161263.1 | - | 1 | 325 | |||
BC100409.1 | - | 1 | 357 | 30919135 | ||
S43865.1 | - | 1 | 354 |