Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI021584.1 | - | 1 | 76 | ||
AK075604.1 | - | 4 | 931 | |||
BC003793.1 | - | 4 | 901 | 3601675 | ||
U28016.1 | - | 4 | 910 |