Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011227.1 | NP_035357.1 | 2 | 640 | ||
GenBank mRNA | AB232614.1 | - | 2 | 640 | ||
AK145566.1 | - | 2 | 817 | |||
AK152454.1 | - | 2 | 766 | |||
AK163743.1 | - | 2 | 780 | |||
AK164541.1 | - | 2 | 780 | |||
BC089570.1 | - | 2 | 765 | 6747570 | ||
BG066967.2 | - | 1 | 534 | |||
X80332.1 | - | 2 | 640 |