Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009008.3 | NP_033034.1 | 3 | 269 | ||
GenBank mRNA | AK007561.1 | - | 3 | 207 | ||
AK011045.1 | - | 3 | 248 | |||
AK088415.1 | - | 3 | 218 | |||
AK088805.1 | - | 3 | 207 | |||
AK144136.1 | - | 3 | 213 | |||
AK171321.1 | - | 3 | 270 | |||
BC005455.1 | - | 3 | 185 | 3495818 | ||
X53247.1 | - | 3 | 167 |