Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297606.1 | NP_001284535.1 | 5 | 588 | ||
NM_001297607.1 | NP_001284536.1 | 5 | 588 | |||
NM_009010.5 | NP_033036.2 | 5 | 588 | |||
GenBank mRNA | AK038300.1 | - | 5 | 565 | ||
AK045224.1 | - | 5 | 578 | |||
AK161875.1 | - | 5 | 565 | |||
AK165467.1 | - | 5 | 589 | |||
BC003846.1 | - | 5 | 576 | |||
BC138684.1 | - | 5 | 548 | 8861705 | ||
BC145372.1 | - | 5 | 548 | 9053930 | ||
X92410.1 | - | 5 | 488 |