Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011274.5 | NP_035404.4 | 11 | 1898 | ||
GenBank mRNA | AF091096.1 | - | 9 | 1420 | ||
AK052034.1 | - | 6 | 1079 | |||
AK133383.1 | - | 11 | 1573 | |||
AK155994.1 | - | 11 | 1871 | |||
AK162285.1 | - | 6 | 1072 | |||
AK165045.1 | - | 4 | 850 | |||
AK166566.1 | - | 11 | 1889 | |||
BC023029.1 | - | 11 | 1558 | 5374731 | ||
BC038523.2 | - | 2 | 195 |