Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159731.1 | NP_001153203.1 | 6, 7 | 974 | ||
NM_009107.3 | NP_033133.1 | 7, 8 | 1430 | |||
GenBank mRNA | BC058401.1 | - | 7, 8 | 1412 | 5707723 | |
G49237.1 | - | 7, 8 | 1162 | |||
M84819.1 | - | 7, 8 | 1075 | |||
S62948.1 | - | 7, 8 | 1274 | |||
S62952.1 | - | 6, 7 | 930 | |||
X66225.1 | - | 7, 8 | 1162 |