Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selplg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009151.3 | NP_033177.3 | 2 | 1134 | ||
GenBank mRNA | AK089188.1 | - | 2 | 1110 | ||
AK089214.1 | - | 2 | 1060 | |||
AK155762.1 | - | 2 | 1067 | |||
AK156933.1 | - | 2 | 1065 | |||
AK158134.1 | - | 2 | 1066 | |||
AK172077.1 | - | 2 | 1068 | |||
BC003874.1 | - | 3 | 1006 | 3495181 | ||
BG070861.2 | - | 1 | 715 | |||
BG083864.2 | - | 1 | 305 | |||
X91144.1 | - | 2 | 923 |