Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011400.3 | NP_035530.2 | 3 | 459 | ||
GenBank mRNA | AK089353.1 | - | 3 | 401 | ||
AK155252.1 | - | 3 | 401 | |||
AK170425.1 | - | 3 | 401 | |||
AK170873.1 | - | 3 | 401 | |||
BC055340.1 | - | 3 | 428 | 6808315 | ||
M22998.1 | - | 3 | 363 | |||
M23384.1 | - | 3 | 438 |