Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145365.3 | NP_663340.1 | 4, 5 | 580 | ||
GenBank mRNA | AF392874.1 | - | 2, 3 | 231 | ||
AK050186.1 | - | 2, 3 | 222 | |||
AK077258.1 | - | 4, 5 | 581 | |||
BC010786.1 | - | 2, 3 | 275 | 4211480 | ||
BC028820.1 | - | 2, 3 | 252 | 5096773 |