Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011528.4 | NP_035658.1 | 7 | 909 | ||
GenBank mRNA | AK003092.1 | - | 7 | 873 | ||
AK142030.1 | - | 7 | 867 | |||
BC003970.1 | - | 7 | 842 | |||
BC004754.1 | - | 7 | 864 | 3584511 | ||
BC094277.1 | - | 7 | 858 | 5008399 | ||
U67611.1 | - | 7 | 852 |