Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291268.1 | NP_001278197.1 | 1 | 249 | ||
NM_009330.3 | NP_033356.2 | 1 | 249 | |||
GenBank mRNA | AB008174.1 | - | 1 | 69 | ||
AB008175.1 | - | 1 | 69 | |||
AB008176.1 | - | 1 | 69 | |||
AB008177.1 | - | 1 | 69 | |||
AB052659.1 | - | 1 | 69 | |||
AK004837.2 | - | 1 | 250 | |||
X55842.1 | - | 1 | 69 |