Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079822.2 | NP_001073290.1 | 10, 11 | 1277 | ||
NM_009332.3 | NP_033358.2 | 9, 10 | 1235 | |||
GenBank mRNA | AF057691.1 | - | 5, 6 | 623 | ||
AJ223069.1 | - | 9, 10 | 1143 | |||
AJ223233.1 | - | 9, 10 | 1058 | |||
AK052721.1 | - | 9, 10 | 1445 | |||
BC128305.1 | - | 10, 11 | 1184 | 40109611 | ||
BC128306.1 | - | 9, 10 | 1142 | 40109613 |