Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098226.3 | NP_001091696.2 | 12 | 1201 | ||
NM_001204156.1 | NP_001191085.1 | 12 | 1524 | |||
NM_011566.4 | NP_035696.3 | 12 | 1365 | |||
GenBank mRNA | AF002670.1 | - | 12 | 1320 | ||
AK087649.1 | - | 12 | 1203 | |||
AK143551.1 | - | 11 | 2237 | |||
BC021364.1 | - | 12 | 1245 | |||
BC027383.1 | - | 3 | 276 | |||
BC039984.1 | - | 3 | 311 | |||
CB182196.1 | - | 1 | 70 | |||
D87963.1 | - | 11 | 1035 | |||
D87964.1 | - | 12 | 1346 | |||
Y10027.1 | - | 11 | 1127 | |||
Y16611.1 | - | 11 | 1118 |