Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Llgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252532.1 | NP_001239461.1 | 16 | 2135 | ||
NM_145438.2 | NP_663413.2 | 16 | 2135 | |||
GenBank mRNA | AK143486.1 | - | 16 | 2123 | ||
AK143798.1 | - | 16 | 2110 | |||
AK167536.1 | - | 16 | 2107 | |||
AY033650.1 | - | 16 | 2085 |