Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177319.1 | NP_001170790.1 | 4 | 426 | ||
NM_001177320.1 | NP_001170791.1 | 4 | 392 | |||
NM_011576.1 | NP_035706.1 | 4 | 589 | |||
GenBank mRNA | AF004833.1 | - | 3 | 358 | ||
AF016313.1 | - | 2 | 233 | |||
AK028356.1 | - | 4 | 589 | |||
AK034752.1 | - | 4 | 393 | |||
AK167522.1 | - | 4 | 451 | |||
BC036146.1 | - | 4 | 426 | 4975683 |