Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198605.3 | NP_941007.1 | 7, 8 | 1178 | ||
GenBank mRNA | AK089223.1 | - | 7, 8 | 1179 | ||
AK166171.1 | - | 7, 8 | 1170 | |||
AK217625.1 | - | 1, 2 | 83 | |||
BC117500.1 | - | 7, 8 | 1170 | 40105921 | ||
BC117501.1 | - | 7, 8 | 1170 | 40105920 |