Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011672.4 | NP_035802.3 | 6 | 590 | ||
GenBank mRNA | AK010461.1 | - | 6 | 559 | ||
AK012543.1 | - | 4 | 534 | |||
AK088823.1 | - | 6 | 559 | |||
AK147138.1 | - | 6 | 573 | |||
AK169062.1 | - | 6 | 572 | |||
BC006630.1 | - | 6 | 547 | 3708320 |