Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009486.3 | NP_033512.1 | 2 | 552 | ||
NM_009487.1 | NP_033513.1 | 2 | 552 | |||
GenBank mRNA | AF011420.1 | - | 2 | 551 | ||
AF011421.1 | - | 2 | 552 | |||
BC145855.1 | - | 2 | 513 | 40131158 |
Set Membership: |
G-protein coupled receptor |