Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oosp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033283.2 | NP_001028455.2 | 5 | 536 | ||
GenBank mRNA | AK143410.1 | - | 5 | 537 | ||
BC094934.1 | - | 5 | 498 | |||
BC141228.1 | - | 5 | 513 | 9055840 | ||
BC145553.1 | - | 4 | 334 |