Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyplal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146106.2 | NP_666218.2 | 3 | 407 | ||
GenBank mRNA | AK148533.1 | - | 3 | 375 | ||
BC027340.1 | - | 3 | 407 | 4023059 |