Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ighv6-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB050074.1 | - | 209 | |||
M59921.1 | - | 199 | ||||
M59922.1 | - | 199 |
Gene Symbol: Igh-VJ558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC049143.1 | - | 2 | 257 | 6474993 |