Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033633.3 | NP_001028805.2 | 3 | 1009 | ||
GenBank mRNA | AK147341.1 | - | 3 | 1010 | ||
BC119586.1 | - | 4 | 576 | 40058865 | ||
BC119587.1 | - | 3 | 518 |